Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOD1
Entrez Gene ID
696222
Gene Name
myogenic differentiation 1
Gene Aliases
EGK_06448
Chromosome Location
Chr.14: 47914251 - 47916888 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015114759.2 XP_014970245.1 - 990 66

View Full Product Details