Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNG11
Entrez Gene ID
700606
Gene Name
G protein subunit gamma 11
Gene Aliases
EGK_13932
Chromosome Location
Chr.3: 112195888 - 112200583 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266194.1 NP_001253123.1 - 409 132
GenBank mRNA JU321237.1 - - 424 132
JU473210.1 - - 173 132
JV043948.1 - - 409 132
JV636910.1 - - 451 132

View Full Product Details