Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPB3
Entrez Gene ID
704576
Gene Name
heat shock protein family B (small) member 3
Gene Aliases
EGK_16480
Chromosome Location
Chr.6: 52783090 - 52783858 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU325371.1 - - 330 169
JV726259.1 - - 278 169

View Full Product Details