Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB25
Entrez Gene ID
707380
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
EGK_18282
Chromosome Location
Chr.7: 126547754 - 126570097 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194209.1 NP_001181138.1 - 179 70
GenBank mRNA JU470765.1 - - 176 70

View Full Product Details