Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDN17
Entrez Gene ID
705582
Gene Name
claudin 17
Gene Aliases
Claudin-17, EGK_13333
Chromosome Location
Chr.3: 16510418 - 16511945 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100305.4 XP_001100305.1 - 502 80

View Full Product Details