Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP10
Entrez Gene ID
700068
Gene Name
bone morphogenetic protein 10
Gene Aliases
EGK_05399
Chromosome Location
Chr.13: 39929939 - 39936222 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001096299.3 XP_001096299.1 - 494 120

View Full Product Details