Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFE
Entrez Gene ID
716913
Gene Name
negative elongation factor complex member E
Gene Aliases
RDBP
Chromosome Location
Chr.4: 137537207 - 137543933 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113911.4 XP_001113911.2 - 1279 124
XM_015136032.2 XP_014991518.2 - 1224 124
XM_015136033.2 XP_014991519.2 - 1380 124
XM_015136034.2 XP_014991520.2 - 1262 124
XM_028847712.1 XP_028703545.1 - 1610 124
GenBank mRNA JU336370.1 - - 1166 124
JU477191.1 - - 1135 124
JV048243.1 - - 1135 124
JV628580.1 - - 1253 124
JV661290.1 - - 1223 124

View Full Product Details