Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XAGE5
Entrez Gene ID
697885
Gene Name
X antigen family member 5
Gene Aliases
-
Chromosome Location
Chr.X: 52573412 - 52580435 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193739.1 NP_001180668.1 - 419 106
XM_028841695.1 XP_028697528.1 - 486 106
XM_028841696.1 XP_028697529.1 - 453 106

Gene Symbol
LOC114669729
Entrez Gene ID
114669729
Gene Name
X antigen family member 5
Gene Aliases
-
Chromosome Location
Chr.X: 52548006 - 52555044 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028841956.1 XP_028697789.1 - 480 106
XM_028841957.1 XP_028697790.1 - 450 106

View Full Product Details