Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRGPRX2
Entrez Gene ID
692078
Gene Name
MAS related GPR family member X2
Gene Aliases
MRGX2
Chromosome Location
Chr.14: 46500778 - 46507502 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040422.1 NP_001035512.1 - 250 116
XM_028832503.1 XP_028688336.1 - 682 116

View Full Product Details