Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIL1
Entrez Gene ID
715283
Gene Name
NFKB inhibitor like 1
Gene Aliases
EGK_14703
Chromosome Location
Chr.4: 137954310 - 137966952 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047144.1 NP_001040609.1 - 56 57
XM_015135923.2 XP_014991409.2 - 183 57
XM_015135924.2 XP_014991410.2 - 171 57
GenBank mRNA JU321545.1 - - 147 57
JV635475.1 - - 134 57
JV669525.1 - - 71 57

View Full Product Details