Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHP2
Entrez Gene ID
701950
Gene Name
NHP2 ribonucleoprotein
Gene Aliases
EGK_17208, NOLA2
Chromosome Location
Chr.6: 175475218 - 175479666 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267011.2 NP_001253940.1 - 293 55
XM_015141552.2 XP_014997038.1 - 313 55
GenBank mRNA JU320751.1 - - 296 55
JU472993.1 - - 313 55
JV045402.1 - - 296 55

View Full Product Details