Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF39
Entrez Gene ID
712215
Gene Name
ring finger protein 39
Gene Aliases
-
Chromosome Location
Chr.4: 139814862 - 139819539 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114946.1 NP_001108418.1 - 582 127

View Full Product Details