Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1C
Entrez Gene ID
717148
Gene Name
CD1c molecule
Gene Aliases
-
Chromosome Location
Chr.1: 92718975 - 92724009 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145533.1 NP_001139005.1 - 606 105
GenBank mRNA AB458513.1 - - 606 105
JU477763.1 - - 890 105
JU477764.1 - - 886 105

View Full Product Details