Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX6
Entrez Gene ID
100286832
Gene Name
glutathione peroxidase 6
Gene Aliases
-
Chromosome Location
Chr.4: 141724093 - 141735212 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159358.3 NP_001152830.2 - 410 81

View Full Product Details