Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
100301991
Gene Name
phosphodiesterase 9A
Gene Aliases
PDE9A2
Chromosome Location
Chr.3: 3888596 - 4009128 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161449.1 NP_001154921.1 - 877 71
XM_028844729.1 XP_028700562.1 - 1232 71
XM_028844730.1 XP_028700563.1 - 965 71
XM_028844731.1 XP_028700564.1 - 554 71
GenBank mRNA FJ914626.1 - - 877 71
JU321678.1 - - 879 71
JU475918.1 - - 870 71
JV584082.1 - - 139 71

View Full Product Details