Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
718468
Gene Name
epididymal sperm binding protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 47978920 - 48002295 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171843.1 NP_001165314.1 - 324 91
GenBank mRNA AF218044.1 - - 324 91

View Full Product Details