Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR16-1
Entrez Gene ID
100315146
Gene Name
microRNA mir-16-1
Gene Aliases
mml-mir-16-1
Chromosome Location
Chr.17: 28503337 - 28503425 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032067.1 - 38 79

View Full Product Details