Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR31
Entrez Gene ID
100315488
Gene Name
microRNA mir-31
Gene Aliases
mir-31, mml-mir-31
Chromosome Location
Chr.15: 62158644 - 62158714 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032003.1 - 37 63

View Full Product Details