Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDI1
Entrez Gene ID
704423
Gene Name
DNA damage inducible 1 homolog 1
Gene Aliases
EGK_06837
Chromosome Location
Chr.14: 97304747 - 97307361 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194140.2 NP_001181069.1 - 2554 95

View Full Product Details