Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX6A2
Entrez Gene ID
713995
Gene Name
cytochrome c oxidase subunit VIa polypeptide 2
Gene Aliases
-
Chromosome Location
Chr.20: 28876911 - 28877540 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194578.2 NP_001181507.1 - 126 64

View Full Product Details