Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP4
Entrez Gene ID
100427947
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
Chromosome Location
Chr.2: 143528019 - 143534275 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194847.3 NP_001181776.1 - 988 93
GenBank mRNA JV722213.1 - - 423 93

View Full Product Details