Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRED1
Entrez Gene ID
100430739
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 14674133 - 14772275 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204360.1 NP_001191289.1 - 328 115
GenBank mRNA HM370981.1 - - 328 115
JU319878.1 - - 121 115
JU319879.1 - - 121 115
JU319880.1 - - 121 115
JV046307.1 - - 264 115
JV535066.1 - - 358 115

View Full Product Details