Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR32
Entrez Gene ID
700923
Gene Name
proline rich 32
Gene Aliases
CXHXorf64, CXorf64
Chromosome Location
Chr.X: 123102213 - 123103905 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193842.2 NP_001180771.2 - 751 110

View Full Product Details