Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2
Entrez Gene ID
710376
Gene Name
BPI fold containing family A member 2
Gene Aliases
-
Chromosome Location
Chr.10: 68380478 - 68391954 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243275.3 NP_001230204.2 - 160 90
XM_028827327.1 XP_028683160.1 - 169 90

View Full Product Details