Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTF3
Entrez Gene ID
703339
Gene Name
basic transcription factor 3
Gene Aliases
EGK_15391
Chromosome Location
Chr.6: 70348846 - 70355981 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261092.2 NP_001248021.1 - 265 88
GenBank mRNA JU471446.1 - - 219 88
JV043938.1 - - 265 88

View Full Product Details