Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CERS2
Entrez Gene ID
716180
Gene Name
ceramide synthase 2
Gene Aliases
EGK_01241, LASS2
Chromosome Location
Chr.1: 100153513 - 100163028 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261642.1 NP_001248571.1 - 594 115
XM_015151027.2 XP_015006513.1 - 665 115
GenBank mRNA JU473393.1 - - 659 115
JV046887.1 - - 594 115
JV528456.1 - - 655 115

View Full Product Details