Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSDHL
Entrez Gene ID
693333
Gene Name
NAD(P) dependent steroid dehydrogenase-like
Gene Aliases
-
Chromosome Location
Chr.X: 148843495 - 148879106 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265710.1 NP_001252639.1 - 908 85
XM_015128441.2 XP_014983927.1 - 1047 85
XM_015128444.2 XP_014983930.1 - 851 85
XM_028841645.1 XP_028697478.1 - 1161 85
XM_028841646.1 XP_028697479.1 - 943 85
XM_028841647.1 XP_028697480.1 - 973 85
GenBank mRNA JU323097.1 - - 958 85
JU475789.1 - - 933 85
JV045287.1 - - 932 85
JV667235.1 - - 908 85

View Full Product Details