Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RARRES2
Entrez Gene ID
704993
Gene Name
retinoic acid receptor responder 2
Gene Aliases
-
Chromosome Location
Chr.3: 176286593 - 176289990 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266683.1 NP_001253612.1 - 582 95
XM_015135190.2 XP_014990676.1 - 1078 95
XM_015135191.2 XP_014990677.1 - 630 95
GenBank mRNA JU473731.1 - - 526 95
JV047825.1 - - 577 95
JV636301.1 - - 582 95

View Full Product Details