Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND5
Entrez Gene ID
711150
Gene Name
BEN domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 174445623 - 175900437 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266827.2 NP_001253756.1 - 368 109
XM_015139078.2 XP_014994564.1 - 610 109
XM_028844223.1 XP_028700056.1 - 954 109
XM_028844227.1 XP_028700060.1 - 918 109
XM_028844233.1 XP_028700066.1 - 956 109
XM_028844260.1 XP_028700093.1 - 793 109
GenBank mRNA JU328464.1 - - 217 109
JV635600.1 - - 370 109

View Full Product Details