Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR7-3
Entrez Gene ID
100315504
Gene Name
microRNA mir-7-3
Gene Aliases
mml-mir-7-3
Chromosome Location
Chr.19: 4706396 - 4706505 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032365.1 - 52 73

View Full Product Details