The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012988.2 | NP_037120.1 | 1 - 2 | 713 | 100 |
GenBank mRNA | AB060652.1 | - | - | 621 | 100 |
AF112455.1 | - | - | 793 | 100 | |
AY572794.1 | - | 1 - 2 | 621 | 100 | |
D78017.1 | - | 1 - 2 | 713 | 100 | |
D78020.1 | - | 1 - 2 | 135 | 100 | |
X13167.1 | - | 1 - 2 | 555 | 100 |