Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1qtnf5
Entrez Gene ID
315598
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
-
Chromosome Location
Chr.8: 48443767 - 48445637 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012123.1 NP_001012123.1 2 - 3 444 109
GenBank mRNA BC089992.1 - 2 - 3 444 109

View Full Product Details