The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013152.2 | NP_037284.2 | 6 - 7 | 950 | 86 |
XM_006252993.2 | XP_006253055.1 | 5 - 6 | 1150 | 86 | |
XM_008771066.1 | XP_008769288.1 | 3 - 4 | 736 | 86 | |
XM_008771067.1 | XP_008769289.1 | 5 - 6 | 1150 | 86 | |
GenBank mRNA | AY168444.1 | - | 3 - 4 | 203 | 86 |
M97380.1 | - | 6 - 7 | 979 | 86 |