The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022210.1 | NP_071546.1 | 4 - 5 | 302 | 71 |
XM_006240248.2 | XP_006240310.1 | 3 - 4 | 501 | 71 | |
GenBank mRNA | D14447.1 | - | 4 - 5 | 302 | 71 |
D14448.1 | - | 3 - 4 | 275 | 71 |