The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012119.1 | NP_001012119.1 | 3 - 4 | 191 | 61 |
XM_006241994.2 | XP_006242056.1 | 3 - 4 | 304 | 61 | |
GenBank mRNA | BC087122.1 | - | 3 - 4 | 191 | 61 |