Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jph3
Entrez Gene ID
307916
Gene Name
junctophilin 3
Gene Aliases
-
Chromosome Location
Chr.19: 54553419 - 54613477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107437.1 NP_001100907.1 2 - 3 1156 61

View Full Product Details