The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012670.1 | NP_036802.1 | 3 - 4 | 358 | 103 |
GenBank mRNA | D90345.1 | - | 3 - 4 | 358 | 103 |