The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012750.1 | NP_036882.1 | 4 - 5 | 917 | 64 |
XM_006252238.2 | XP_006252300.1 | 3 - 4 | 1686 | 64 | |
GenBank mRNA | AF003825.1 | - | - | 1467 | 64 |
AF005226.1 | - | 4 - 5 | 797 | 64 | |
U97143.1 | - | 4 - 5 | 917 | 64 |