Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo1
Entrez Gene ID
25256
Gene Name
flavin containing monooxygenase 1
Gene Aliases
RFMO1A
Chromosome Location
Chr.13: 80712880 - 80745095 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012792.1 NP_036924.1 2 - 3 173 60
XM_006250145.2 XP_006250207.1 2 - 3 260 60
XM_006250146.2 XP_006250208.1 2 - 3 157 60
GenBank mRNA BC061567.1 - 2 - 3 166 60
FQ218919.1 - 2 - 3 381 60
M84719.1 - 2 - 3 173 60

View Full Product Details