Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gp5
Entrez Gene ID
25259
Gene Name
glycoprotein V (platelet)
Gene Aliases
PLGPV
Chromosome Location
Chr.11: 74014983 - 74018075 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012795.2 NP_036927.1 2 - 2 1481 139
XM_006248483.2 XP_006248545.1 2 - 2 1767 139

Gene Symbol
LOC100911551
Entrez Gene ID
100911551
Gene Name
platelet glycoprotein V-like
Gene Aliases
-
Chromosome Location
Chr.11: 73015358 - 73017248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751076.3 XP_003751124.2 1 - 1 1479 139

View Full Product Details