The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012900.1 | NP_037032.1 | 10 - 11 | 828 | 63 |
GenBank mRNA | U35097.1 | - | 10 - 11 | 828 | 63 |
Z50083.1 | - | 10 - 11 | 786 | 63 | |
Z50084.1 | - | 6 - 7 | 495 | 63 |