Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctse
Entrez Gene ID
25424
Gene Name
cathepsin E
Gene Aliases
CEA, CEB, Ctsea
Chromosome Location
Chr.13: 48426833 - 48449493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012938.1 NP_037070.1 1 - 2 88 134
XM_006249748.2 XP_006249810.1 1 - 2 293 134
GenBank mRNA BC062002.1 - 1 - 2 98 134
D38104.1 - 1 - 2 88 134
D45187.1 - 1 - 2 230 134

View Full Product Details