Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gys2
Entrez Gene ID
25623
Gene Name
glycogen synthase 2
Gene Aliases
GLYSN
Chromosome Location
Chr.4: 176638633 - 176679621 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF346902.1 - 14 - 15 1807 100
J05446.1 - 14 - 15 1849 100

View Full Product Details