The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013111.3 | NP_037243.2 | - | 961 | 76 |
XM_008768943.1 | XP_008767165.1 | 3 - 4 | 1233 | 76 | |
XM_008768944.1 | XP_008767166.1 | 3 - 4 | 1233 | 76 | |
GenBank mRNA | AF245000.1 | - | - | 901 | 76 |
D67087.1 | - | 3 - 4 | 691 | 76 | |
L10152.1 | - | 3 - 4 | 685 | 76 | |
U70476.1 | - | 3 - 4 | 770 | 76 |