Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cartpt
Entrez Gene ID
29131
Gene Name
CART prepropeptide
Gene Aliases
Cart
Chromosome Location
Chr.2: 30125249 - 30127693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017110.1 NP_058806.1 1 - 2 174 56
GenBank mRNA U10071.1 - 1 - 2 174 56

View Full Product Details