The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017189.2 | NP_058885.1 | 7 - 8 | 815 | 61 |
XM_006246676.2 | XP_006246738.1 | 7 - 8 | 893 | 61 | |
GenBank mRNA | AF230645.1 | - | 6 - 7 | 901 | 61 |
BC088099.1 | - | 7 - 8 | 695 | 61 | |
FQ218583.1 | - | 5 - 6 | 521 | 61 | |
J02762.1 | - | 7 - 8 | 815 | 61 | |
M16347.1 | - | 7 - 8 | 61 | ||
X07636.1 | - | 6 - 7 | 718 | 61 |