Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gsk3a
Entrez Gene ID
50686
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
-
Chromosome Location
Chr.1: 82097244 - 82108203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017344.1 NP_059040.1 8 - 9 1225 121
XM_008758972.1 XP_008757194.1 8 - 9 1234 121
GenBank mRNA X53427.1 - 8 - 9 1225 121

View Full Product Details