Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk6
Entrez Gene ID
29245
Gene Name
kallikrein related-peptidase 6
Gene Aliases
Prss9
Chromosome Location
Chr.1: 99762221 - 99769496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019175.1 NP_062048.1 6 - 7 788 68
XM_006228982.2 XP_006229044.1 6 - 7 895 68
GenBank mRNA AF016269.1 - 6 - 7 788 68

View Full Product Details