The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083911.1 | NP_001077380.1 | 3 - 4 | 569 | 131 |
NM_019192.2 | NP_062065.2 | 3 - 4 | 497 | 131 | |
GenBank mRNA | BC059137.1 | - | 3 - 4 | 498 | 131 |
BC072539.1 | - | 3 - 4 | 486 | 131 | |
D25221.1 | - | 3 - 4 | 478 | 131 | |
FQ218512.1 | - | 1 - 2 | 126 | 131 | |
M63574.1 | - | - | 457 | 131 |