The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006239476.2 | XP_006239538.1 | 5 - 6 | 1182 | 58 |
XM_006239479.2 | XP_006239541.1 | 5 - 6 | 1182 | 58 | |
XM_008764375.1 | XP_008762597.1 | 5 - 6 | 1182 | 58 | |
XM_008764376.1 | XP_008762598.1 | 5 - 6 | 1182 | 58 | |
XM_008764377.1 | XP_008762599.1 | 5 - 6 | 1182 | 58 | |
XM_008764378.1 | XP_008762600.1 | 5 - 6 | 1182 | 58 | |
XM_008764379.1 | XP_008762601.1 | 5 - 6 | 1182 | 58 | |
XM_008764380.1 | XP_008762602.1 | 5 - 6 | 1182 | 58 | |
GenBank mRNA | U46007.1 | - | 5 - 6 | 1005 | 58 |